Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
citrullinated RGG Rabbit anti-Human, Unconjugated, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$182.31 - $457.74
Specifications
Antigen | citrullinated RGG |
---|---|
Applications | ELISA, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABS224425MI
|
MilliporeSigma
ABS224425UG |
25 μg |
Each for $182.31
|
|
|||||
ABS224410MI
|
MilliporeSigma
ABS2244100UG |
100 μg |
Each for $457.74
|
|
|||||
Description
citrullinated RGG Polyclonal antibody specifically detects citrullinated RGG in Human samples. It is validated for ELISA, Western Blotting.Specifications
citrullinated RGG | |
Polyclonal | |
Purified | |
Inflammation and Immunology | |
Deaminated RGG | |
IgG | |
Affinity Purified | |
This rabbit polyclonal antibody specifically detects citrullinated RGG motifs in proteins. |
ELISA, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
A chemically synthesized citrullinated peptide (CGGCitGGGCitGG). | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
Multiple bands based on multiple protein citrullination. Uncharacterized bands may be observed in some lysate(s). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title