Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CK 666
GREENER_CHOICE

Catalog No. p-4816738
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Arp2/3 inhibitor; inhibits actin polymerization

Inhibitor of the Arp2/3 complex; inhibits actin polymerization (IC50 = 4 μM).

Chemical Identifiers

CAS 442633-00-3
Molecular Formula C18H17FN2O

Specifications

Quantity 10 mg
Formula Weight 296.34
Purity >99%
Molecular Weight (g/mol) 296.345
Chemical Name or Material CK 666
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.