Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CK1 delta Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518509
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518509 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518509 Supplier Invitrogen™ Supplier No. PA518509
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

CSNK1D (CK1 delta), a serine/threonine kinase, plays a critical role in Wnt/beta-catenin signaling pathway. This pathway is important in both development and cancer. The encoded protein may also be involved in the regulation of apoptosis, circadian rhythm, microtubule dynamics, chromosome segregation, and p53-mediated effects on growth. The encoded protein is highly similar to the mouse and rat CK1 delta homologs. Three transcript variants encoding different isoforms have been found for CK1 delta.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CK1 delta
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene CSNK1D
Gene Accession No. P48730, Q9DC28
Gene Alias 1200006A05Rik; AA409348; ASPS; casein kinase 1; casein kinase 1 delta; casein kinase 1, delta; casein kinase I isoform delta; CK1d; CK1delta; CKId; CKIdelta; CKI-delta; CSNK1D; D930010H05Rik; FASPS2; HCKID; HGNC:2452; Tau-protein kinase CSNK1D
Gene Symbols CSNK1D
Host Species Goat
Immunogen Synthetic peptide sequence (DREERLRHSRNPATR) corresponding to the internal amino acids of CSNK1D (aa 310-325).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104318, 1453
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.