Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CKAP1 Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB065183 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB065183 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB065183 Supplier Novus Biologicals Supplier No. NBP300264G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CKAP1 Polyclonal specifically detects CKAP1 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen CKAP1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias CG22Tubulin-specific chaperone B, CKAP1MGC14625, CKAPI, cytoskeleton associated protein 1, Cytoskeleton-associated protein 1Cytoskeleton-associated protein CKAPI, tubulin folding cofactor B, tubulin-folding cofactor B
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant CKAP1 (Q99426; Met1-Ile244).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1155
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.