Learn More
Description
Specifications
Specifications
| Antigen | CKAP1 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | HRP |
| Dilution | Western Blot, ELISA |
| Formulation | PBS |
| Gene Alias | CG22Tubulin-specific chaperone B, CKAP1MGC14625, CKAPI, cytoskeleton associated protein 1, Cytoskeleton-associated protein 1Cytoskeleton-associated protein CKAPI, tubulin folding cofactor B, tubulin-folding cofactor B |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant CKAP1 (Q99426; Met1-Ile244). |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
