Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CKAP2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP121391 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP121391 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP121391 Supplier Novus Biologicals Supplier No. NBP121391
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CKAP2 Polyclonal specifically detects CKAP2 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen CKAP2
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_001091995.1
Gene Alias CTCL tumor antigen se20-10, cytoskeleton associated protein 2, cytoskeleton-associated protein 2, LB1DKFZp686L1238, se20-10, TMAPFLJ10749, Tumor- and microtubule-associated protein, tumor-associated microtubule-associated protein
Gene Symbols CKAP2
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 633 and 683 of human cytoskeleton associated protein 2 using the numbering given in entry NP_001091995.1 (GeneID 26586).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 26586
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.