Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CKI gamma 3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15757320 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15757320 20 μL
NBP157573 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15757320 Supplier Novus Biologicals Supplier No. NBP15757320UL

Rabbit Polyclonal Antibody

CKI gamma 3 Polyclonal specifically detects CKI gamma 3 in Human samples. It is validated for Western Blot.

Specifications

Antigen CKI gamma 3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9Y6M4
Gene Alias casein kinase 1, gamma 3, casein kinase I isoform gamma-3, CKI-gamma 3, EC 2.7.11, EC 2.7.11.1
Gene Symbols CSNK1G3
Host Species Rabbit
Immunogen Synthetic peptides corresponding to CSNK1G3(casein kinase 1, gamma 3) The peptide sequence was selected from the middle region of CSNK1G3. Peptide sequence LEALGHMFMYFLRGSLPWQGLKADTLKERYQKIGDTKRATPIEVLCENFP.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 1456
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.