Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CKII alpha prime polypeptide Antibody, DyLight 680, Novus Biologicals™
SDP

Catalog No. NB086065 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB086065 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB086065 Supplier Novus Biologicals Supplier No. NBP299686FR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CKII alpha prime polypeptide Polyclonal specifically detects CKII alpha prime polypeptide in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CKII alpha prime polypeptide
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 680
Formulation 50 mM Sodium Borate
Gene Alias casein kinase 2, alpha prime polypeptide, casein kinase II subunit alpha', CK II alpha', CK2A2, CSNK2A1, EC 2.7.11, EC 2.7.11.1, FLJ43934
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant CKII alpha prime polypeptide (NP_001887.1; Met1-Arg350).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 1459
Test Specificity Human CSNK2A2
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.