Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CKIP-1 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB358670 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB358670 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB358670 Supplier Novus Biologicals Supplier No. NBP176238PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CKIP-1 Polyclonal antibody specifically detects CKIP-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen CKIP-1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias 2810052M02Rik, Casein kinase 2-interacting protein 1, C-Jun-binding protein, CK2 interacting protein 1; HQ0024c protein, CK2-interacting protein 1, CKIP1, CKIP-1PH domain-containing family O member 1, JBP, OC120, Osteoclast maturation-associated gene 120 protein, pleckstrin homology domain containing, family O member 1, pleckstrin homology domain-containing family O member 1
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid synthetic peptide from near the carboxy terminus of human CKIP1. The immunogen is located within the last 50 amino acids of CKIP1. Amino Acid Squence: TPDSHLRQTTPHSQYKKS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 51177
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.