Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Cl-4AS-1
GREENER_CHOICE

Catalog No. 381210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-1210 10 mg
38-125-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 38-1210 Supplier Tocris Bioscience™ Supplier No. 3812/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Steroidal androgen receptor agonist

Potent steroidal androgen receptor agonist (IC50 = 12 nM). Mimics the action of 5α-dihydrotestosterone (DHT). Transactivates the mouse mammary tumor virus (MMTV) promoter; represses MMP1 promoter activity. Inhibits 5α-reductase type I and II (IC50 values are 6 and 10 nM respectively).

Chemical Identifiers

CAS 188589-66-4
Molecular Formula C26H33ClN2O2

Specifications

Quantity 10 mg
Formula Weight 441.01
Purity >99%
Molecular Weight (g/mol) 441.012
Chemical Name or Material Cl-4AS-1
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.