Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLASP2 Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NBP12395549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP12395549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP12395549 Supplier Novus Biologicals Supplier No. NBP121395JF549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CLASP2 Polyclonal specifically detects CLASP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CLASP2
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CLIP-associating protein 2, cytoplasmic linker associated protein 2, Cytoplasmic linker-associated protein 2, hOrbit2, KIAA0627, multiple asters (Mast)-like homolog 2, Protein Orbit homolog 2
Gene Symbols CLASP2
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 275 and 325 of human cytoplasmic linker associated protein 2 using the numbering given in entry BAG11221.1 (GeneID 23122).
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 23122
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.