Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Claspin Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB292345 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292345 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292345 Supplier Novus Biologicals Supplier No. NB100246APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Claspin Polyclonal specifically detects Claspin in Human samples. It is validated for Immunoprecipitation (Negative), Western Blot.

Specifications

Antigen Claspin
Applications Immunoprecipitation (Negative), Western Blot
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunoprecipitation (Negative)
Formulation PBS
Gene Alias claspin, claspin homolog, claspin homolog (Xenopus laevis), hClaspin, MGC131612, MGC131613, MGC131615
Host Species Rabbit
Immunogen Maps to a region between residues 450 and 500 of human Claspin using the numbering given in entry AAG24515.1 (GeneID 63967).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 63967
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.