Learn More
Antistat Class 100 Cleanroom LDPE Bag
Class 100 Cleanroom LDPE bags made from ultra clean low-density polyethylene, produced within a Class 100 / ISO 5 cleanroom.
Supplier: Antistat 6000173
Description
- Manufactured in Class 100 cleanroom
- Heat sealable
- Ultra low outgassing material
- Supplied double-bagged
- Thickness: 4.0 mil
- Suitable for automotive, biotechnology, electronics, medical devices, consumer goods, life science and healthcare.