Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clathrin Heavy Chain 1/CHC17 Antibody (CLTC/1431), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP25430A750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP25430A750 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP25430A750 Supplier Novus Biologicals Supplier No. NBP254300AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Clathrin Heavy Chain 1/CHC17 Monoclonal specifically detects Clathrin Heavy Chain 1/CHC17 in Human samples. It is validated for ELISA.

Specifications

Antigen Clathrin Heavy Chain 1/CHC17
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone CLTC/1431
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CHC, CHC17, clathrin heavy chain 1, Clathrin heavy chain on chromosome 17, clathrin, heavy chain, clathrin, heavy chain (Hc), clathrin, heavy polypeptide (Hc), clathrin, heavy polypeptide-like 2, CLH17, CLH-17, CLTCL2, Hc, KIAA0034
Gene Symbols CLTC
Host Species Mouse
Immunogen Purified recombinant N-terminal fragment of human Clathrin Heavy Chain (exact sequence is proprietary)
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 1213
Test Specificity Recognizes protein of 192kDa, which is identified as Clathrin Heavy Chain. Clathrin is composed of three heavy chains and three light chains, which associate non-covalently to form a triskelion structure. Clathrin heavy chain (HC) is composed of a terminal globular domain, a distal segment and a proximal segment containing a light chain-binding site. The proximal segment of the Clathrin HC protein is essential for interactions between Clathrin heavy chains and light chains, which result in the formation of the triskelion structure.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.