Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clathrin Heavy Chain 1/CHC17 Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB084683 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB084683 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB084683 Supplier Novus Biologicals Supplier No. NBP299308H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Clathrin Heavy Chain 1/CHC17 Polyclonal specifically detects Clathrin Heavy Chain 1/CHC17 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Clathrin Heavy Chain 1/CHC17
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias CHC, CHC17, clathrin heavy chain 1, Clathrin heavy chain on chromosome 17, clathrin, heavy chain, clathrin, heavy chain (Hc), clathrin, heavy polypeptide (Hc), clathrin, heavy polypeptide-like 2, CLH17, CLH-17, CLTCL2, Hc, KIAA0034
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human Clathrin Heavy Chain 1/CHC17.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Membrane Trafficking and Chaperones, Membrane Vesicle Markers
Primary or Secondary Primary
Gene ID (Entrez) 1213
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.