Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clathrin Heavy Chain 1/CHC17 Antibody (TD.1), DyLight 550
SDP

Catalog No. NB12011331R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB12011331R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB12011331R Supplier Novus Biologicals Supplier No. NB12011331R

Mouse Monoclonal Antibody

Clathrin Heavy Chain 1/CHC17 Monoclonal specifically detects Clathrin Heavy Chain 1/CHC17 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot.

Specifications

Antigen Clathrin Heavy Chain 1/CHC17
Applications Western Blot
Classification Monoclonal
Clone TD.1
Conjugate DyLight 550
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CHC, CHC17, clathrin heavy chain 1, Clathrin heavy chain on chromosome 17, clathrin, heavy chain, clathrin, heavy chain (Hc), clathrin, heavy polypeptide (Hc), clathrin, heavy polypeptide-like 2, CLH17, CLH-17, CLTCL2, Hc, KIAA0034
Gene Symbols CLTC
Host Species Mouse
Immunogen Full length native protein (purified) (Cow).
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Membrane Trafficking and Chaperones, Membrane Vesicle Markers
Primary or Secondary Primary
Gene ID (Entrez) 1213
Test Specificity Monoclonal Anti-Clathrin Heavy Chain reacts specifically with the clathrin heavy chain (CHC17), and does not cross-react with the muscle isoform CHC22. The epitope recognized by the antibody resides within amino acids 207-264 of bovine clathrin heavy chains. It is reported to be non-reactive with native clathrin in immunocytochemistry and immunoprecipitation (but some staining is seen in Rat1 cells, following 4% paraformaldehyde-0.5% Triton X-100 fixation).
Target Species Human, Mouse, Rat, Bovine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.