Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clathrin Heavy Chain 1/CHC17 Antibody (X22), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB300613APC Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300613APC 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300613APC Supplier Novus Biologicals Supplier No. NB300613APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Clathrin Heavy Chain 1/CHC17 Monoclonal specifically detects Clathrin Heavy Chain 1/CHC17 in Human, Mouse, Rat, Bovine, Hamster, Primate samples. It is validated for Flow Cytometry.

Specifications

Antigen Clathrin Heavy Chain 1/CHC17
Applications Flow Cytometry
Classification Monoclonal
Clone X22
Conjugate APC
Dilution Flow Cytometry
Gene Accession No. Q00610
Gene Alias CHC, CHC17, clathrin heavy chain 1, Clathrin heavy chain on chromosome 17, clathrin, heavy chain, clathrin, heavy chain (Hc), clathrin, heavy polypeptide (Hc), clathrin, heavy polypeptide-like 2, CLH17, CLH-17, CLTCL2, Hc, KIAA0034
Gene Symbols CLTC
Host Species Mouse
Immunogen Purified human brain clathrin heavy chain.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Membrane Trafficking and Chaperones, Membrane Vesicle Markers
Primary or Secondary Primary
Gene ID (Entrez) 1213
Test Specificity Detects clathrin heavy chain.
Target Species Human, Mouse, Rat, Bovine, Hamster, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.