Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Clathrin Heavy Chain Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143896
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143896 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143896 Supplier Invitrogen™ Supplier No. PA5143896
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human HEK293 whole cell, human Raji whole cell, rat brain tissue, rat kidney tissue, mouse brain tissue, mouse kidney tissue, mouse ANA-1 whole cell. IHC: human lung cancer tissue, mouse intestine tissue, rat intestine tissue. ICC/IF: U20S cell. Flow: U87 cell, HEPA1-6 cell, C6 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Clathrin is the major protein of the polyhedral coat of coated pits and vesicles. Two different adapter protein complexes link the clathrin lattice either to the plasma membrane or to the trans-Golgi network.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Clathrin Heavy Chain
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.01mg sodium azide
Gene CLTC
Gene Accession No. P11442, P53675, Q00610, Q68FD5
Gene Alias 3110065L21Rik; CHC; CHC17; CHC22; clathrin heavy chain; clathrin heavy chain 1; Clathrin heavy chain 2; clathrin heavy chain like 1; clathrin heavy chain on chromosome 17; Clathrin heavy chain on chromosome 22; clathrin, heavy chain (Hc); clathrin, heavy chain-like 1; clathrin, heavy polypeptide (Hc); Clathrin, heavy polypeptide D; clathrin, heavy polypeptide-like 1; clathrin, heavy polypeptide-like 2; CLH17; CLH-17; CLH22; CLH-22; Cltc; CLTCL; CLTCL1; CLTCL2; CLTD; Hc; KIAA0034; R74732
Gene Symbols CLTC, CLTCL1
Host Species Rabbit
Immunogen E.coli-derived human Clathrin heavy chain/CLTC recombinant protein (Position: R967-Q1668).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1213, 54241, 67300, 8218
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.