Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clathrin interactor 1 Antibody (OTI8C1), PerCP, Novus Biologicals™
SDP

Catalog No. NB288829 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288829 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288829 Supplier Novus Biologicals Supplier No. NBP271491PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Clathrin interactor 1 Monoclonal antibody specifically detects Clathrin interactor 1 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Clathrin interactor 1
Applications Western Blot
Classification Monoclonal
Clone OTI8C1
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias clathrin interacting protein localized in the trans-Golgi region, clathrin interactor 1, Clathrin-interacting protein localized in the trans-Golgi region, CLINT, ENTHClint, enthoprotin, EPN4Enthoprotin, EPNRepsin 4, Epsin-4, EpsinR, Epsin-related protein, FLJ46753, KIAA0171epsinR
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-290 of human CLINT1(NP_055481) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9685
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.