Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clathrin light chain + heavy chain Antibody, Novus Biologicals™
SDP

Catalog No. NBP105035 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP105035 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP105035 Supplier Novus Biologicals Supplier No. NBP105035
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 3 publications

Clathrin light chain + heavy chain Polyclonal specifically detects Clathrin light chain + heavy chain in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen Clathrin light chain + heavy chain
Applications Western Blot, Immunohistochemistry (Paraffin), ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.1-0.3 ug/ml, Immunohistochemistry-Paraffin 2.5 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_001824.1
Gene Alias clathrin light chain A, clathrin, light chain A, LCA, light polypeptide (Lca)
Gene Symbols CLTA
Host Species Goat
Immunogen Peptide with sequence CDFNPKSSKQAKD corresponding to internal region according to NP_009027.1, NP_001070145.1, NP_001825.1, NP_009028.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Membrane Trafficking and Chaperones, Membrane Vesicle Markers
Primary or Secondary Primary
Gene ID (Entrez) 1211
Test Specificity This antibody is expected to recognize all reported isoforms of light chains a and b.
Target Species Human, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.