Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Clathrin light chain Mouse anti-Human, Clone-SPM174, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP254435200UG
Description
Human chromosome location: 9p13.3 IHC-P: Staining of formalin-fixed tissues requires boiling tissue sections in 10mM Citrate Buffer pH 6.0 for 10-20 min followed by cooling at RT for 20 minutesSpecifications
Clathrin light chain | |
Monoclonal | |
1 mg/mL | |
Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, CyTOF-ready | |
clathrin light chain A, clathrin, light chain A, LCA, light polypeptide (Lca) | |
Mouse | |
37 kDa | |
200 μg | |
Membrane Trafficking and Chaperones, Membrane Vesicle Markers | |
1211 | |
Store at -20 to -80C. Avoid freeze-thaw cycles. |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
SPM174 | |
Unconjugated | |
PBS with No Preservative | |
CLTA | |
Purified recombinant N-terminal fragment of human Clathrin Light Chain | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction