Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Clathrin light chain Mouse anti-Human, Clone-SPM174, Novus Biologicals™

Mouse Monoclonal Antibody
$646.00
Specifications
Antigen | Clathrin light chain |
---|---|
Clone | SPM174 |
Concentration | 1 mg/mL |
Dilution | Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB254435100
![]() |
Novus Biologicals
NBP254435100UG |
100 μg |
Each for $646.00
|
|
|||||
NB254435200
![]() |
Novus Biologicals
NBP254435200UG |
200 μg | N/A | N/A | N/A | ||||
Description
Clathrin light chain Monoclonal specifically detects Clathrin light chain in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
Clathrin light chain | |
1 mg/mL | |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
Unconjugated | |
RUO | |
Human | |
clathrin light chain A, clathrin, light chain A, LCA, light polypeptide (Lca) | |
CLTA | |
IgG2b κ | |
Protein A or G purified | |
37 kDa |
SPM174 | |
Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, CyTOF-ready | |
Monoclonal | |
Mouse | |
Membrane Trafficking and Chaperones, Membrane Vesicle Markers | |
PBS with No Preservative | |
1211 | |
Purified recombinant N-terminal fragment of human Clathrin Light Chain | |
Primary | |
Store at -20 to -80C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title