Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clathrin light chain Mouse, DyLight 488, Clone: CON.1, Novus Biologicals™
SDP

Catalog No. NB6001520G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6001520G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB6001520G Supplier Novus Biologicals Supplier No. NB6001520G

Mouse Monoclonal Antibody

Clathrin light chain Monoclonal specifically detects Clathrin light chain in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot.

Specifications

Antigen Clathrin light chain
Applications Western Blot
Classification Monoclonal
Clone CON.1
Conjugate DyLight 488
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias clathrin light chain A, clathrin, light chain A, LCA, light polypeptide (Lca)
Gene Symbols CLTA
Host Species Mouse
Immunogen Synthetic peptide, corresponding to amino acids 23-44 of Cow Clathrin Light Chain.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Membrane Trafficking and Chaperones, Membrane Vesicle Markers
Primary or Secondary Primary
Gene ID (Entrez) 1211
Test Specificity Monoclonal Anti-Clathrin Light Chain reacts specifically with the clathrin light chain. The epitope recognized by the antibody resides within amino acids 23-44 of bovine clathrin light chains, LCa and LCb. This region is not involved in heavy chain binding, and is completely conserved in all mammals. The product is useful in immunoblotting 2-4 (two doublets at 25-36 kDa, a doublet at approximately 56 kDa, and possibly also additional lower and higher molecular weight bands), immunoprecipitation, immunocytochemistry (4% paraformaldehyde- 0.5% Triton X-100 fixation), and immunoaffinity purification. Cross-reactivity has been observed with human, bovine, rat, mouse, and yeast.
Target Species Human, Mouse, Rat, Bovine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.