Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Claudin-1 Antibody (421203), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. NB312390 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB312390 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB312390 Supplier Novus Biologicals Supplier No. FAB4618PECY55
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Claudin-1 Monoclonal specifically detects Claudin-1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Claudin-1
Applications Flow Cytometry
Classification Monoclonal
Clone 421203
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS
Gene Alias claudin 1, claudin-1, CLD1, ILVASCSenescence-associated epithelial membrane protein, SEMP1senescence-associated epithelial membrane protein 1
Host Species Rat
Immunogen CHO Chinese hamster ovary cell line transfected with human Claudin-1, Met1-Val211, Accession # O95832
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 9076
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.