Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Claudin-1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP177036 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP177036 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP177036 Supplier Novus Biologicals Supplier No. NBP177036
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 8 publications

Claudin-1 Polyclonal specifically detects Claudin-1 in Human, Mouse, Rat, Porcine, Canine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunofluorescence, Immunocytochemistry.

Specifications

Antigen Claudin-1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 5-20 ug/mL
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. O95832
Gene Alias claudin 1, claudin-1, CLD1, ILVASCSenescence-associated epithelial membrane protein, SEMP1senescence-associated epithelial membrane protein 1
Gene Symbols CLDN1
Host Species Rabbit
Immunogen Antibody was raised against a 20 amino acid synthetic peptide near the carboxy terminus of human CLDN1. The immunogen is located within the last 50 amino acids of CLDN1. Amino Acid Squence: PRKTTSYPTPRPYPKPAPS
Molecular Weight of Antigen 23 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 9076
Target Species Human, Mouse, Rat, Pig
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.