Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Claudin-1 Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB214884 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB214884 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB214884 Supplier Novus Biologicals Supplier No. NBP177036R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Claudin-1 Polyclonal antibody specifically detects Claudin-1 in Human, Mouse, Rat, Porcine, Canine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence

Specifications

Antigen Claudin-1
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias claudin 1, claudin-1, CLD1, ILVASCSenescence-associated epithelial membrane protein, SEMP1senescence-associated epithelial membrane protein 1
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 20 amino acids near the carboxy terminus of human Claudin-1. The immunogen is located within the last 50 amino acids of Claudin-1. Amino Acid Sequence: PRKTTSYPTPRPYPKPAPS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 9076
Target Species Human, Mouse, Rat, Pig, Canine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.