Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Claudin-1 Antibody, Novus Biologicals™
SDP

Catalog No. NB018074 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018074 0.1 mL
NB12015098 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018074 Supplier Novus Biologicals Supplier No. NB120150980.1ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Claudin-1 Polyclonal specifically detects Claudin-1 in Human, Mouse, Rat, Canine, Primate samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Claudin-1
Applications Immunohistochemistry (Paraffin), Immunohistochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:200
Formulation Citrate, pH 6.0, containing less than 0.1% sodium azide with Sodium Azide
Gene Alias claudin 1, claudin-1, CLD1, ILVASCSenescence-associated epithelial membrane protein, SEMP1senescence-associated epithelial membrane protein 1
Gene Symbols CLDN1
Host Species Rabbit
Immunogen Synthetic peptide (Human) (C terminal).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 9076
Test Specificity Claudin 1
Target Species Rat, Canine, Mouse, Primate, Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.