Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Claudin-10b Antibody (394513), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB312528 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB312528 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB312528 Supplier Novus Biologicals Supplier No. FAB4279APCCY7

Mouse Monoclonal Antibody

Claudin-10b Monoclonal antibody specifically detects Claudin-10b in Human samples. It is validated for Flow Cytometry

Specifications

Antigen Claudin-10b
Applications Flow Cytometry
Classification Monoclonal
Clone 394513
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias CLDN10;CPETRL3;HELIX;OSPL;OSP-L
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Claudin-10 b Isoform, Met1-Val228, Accession # P78369
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9071
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.