Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Claudin-11 Antibody (398205R), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. NB312522 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB312522 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB312522 Supplier Novus Biologicals Supplier No. FAB4280RPECY55
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Claudin-11 Monoclonal antibody specifically detects Claudin-11 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen Claudin-11
Applications Flow Cytometry
Classification Monoclonal
Clone 398205R
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS
Gene Alias claudin 11, claudin-11, Oligodendrocyte-specific protein, OSPOTMoligodendrocyte transmembrane protein
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Claudin-11, Met1-Val207, Accession # O75508
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5010
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.