Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Claudin-18 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.00 - $487.50

Specifications

Antigen Claudin-18
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP15925020
SDP
View Documents
Novus Biologicals
NBP15925020UL
20 μL
Each for $206.00
Only null left
Add to Cart
 
NBP159250
SDP
View Documents
Novus Biologicals
NBP159250
100 μL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

Claudin-18 Polyclonal specifically detects Claudin-18 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Claudin-18
Polyclonal
Rabbit
P56856
CLDN18
IgG
Western Blot
Unconjugated
RUO
51208
Synthetic peptides corresponding to CLDN18(claudin 18) The peptide sequence was selected from the middle region of CLDN18. Peptide sequence YHASGHSVAYKPGGFKASTGFGSNTKNKKIYDGGARTEDEVQSYPSKHDY.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.