Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Claudin-18 Mouse anti-Human, Clone: 2, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314642100UL
This item is not returnable.
View return policy
Description
Claudin-18 Monoclonal antibody specifically detects Claudin-18 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Claudin-18 | |
| Monoclonal | |
| Unconjugated | |
| 0.2μm filtered solution in PBS without Preservative | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| 2 | |
| Immunohistochemistry-Paraffin 1:1000-1:5000 | |
| claudin 18, Claudin 18.2, claudin-18, DKFZp564B2062, SFTA5, SFTPJ, surfactant associated 5, surfactant associated protein J, surfactant, pulmonary associated protein J | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the human Claudin 18 | |
| 100 μg | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 51208 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction