Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Claudin-18 Mouse anti-Human, Clone: 2, Novus Biologicals™

Mouse Monoclonal Antibody
$593.00
Specifications
Antigen | Claudin-18 |
---|---|
Clone | 2 |
Dilution | Immunohistochemistry-Paraffin 1:1000-1:5000 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
Claudin-18 Monoclonal antibody specifically detects Claudin-18 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Claudin-18 | |
Immunohistochemistry-Paraffin 1:1000-1:5000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
claudin 18, Claudin 18.2, claudin-18, DKFZp564B2062, SFTA5, SFTPJ, surfactant associated 5, surfactant associated protein J, surfactant, pulmonary associated protein J | |
Produced in rabbits immunized with a synthetic peptide corresponding to the human Claudin 18 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
2 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cell Biology, Cytoskeleton Markers, Signal Transduction | |
0.2μm filtered solution in PBS without Preservative | |
51208 | |
IgG2a | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title