Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Claudin-3 Antibody (385021), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. NB312382 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB312382 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB312382 Supplier Novus Biologicals Supplier No. FAB4620PECY55
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Claudin-3 Monoclonal specifically detects Claudin-3 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Claudin-3
Applications Flow Cytometry
Classification Monoclonal
Clone 385021
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS
Gene Alias C7orf1, claudin 3, claudin-3, Clostridium perfringens enterotoxin receptor 2, CPE-R 2, CPE-R2, CPETR2CPE-receptor 2, HRVP1, Rat ventral prostate.1 protein homolog, RVP1, ventral prostate.1 protein homolog, ventral prostate.1-like protein
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human Claudin-3, Met1-Val220, Accession # O15551
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 1365
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.