Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Claudin-3 Antibody (OTI1E7), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB289583 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289583 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289583 Supplier Novus Biologicals Supplier No. NBP270445APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Claudin-3 Monoclonal antibody specifically detects Claudin-3 in Human, Mouse, Rat, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen Claudin-3
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1E7
Conjugate APC
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias C7orf1, claudin 3, claudin-3, Clostridium perfringens enterotoxin receptor 2, CPE-R 2, CPE-R2, CPETR2CPE-receptor 2, HRVP1, Rat ventral prostate.1 protein homolog, RVP1, ventral prostate.1 protein homolog, ventral prostate.1-like protein
Host Species Mouse
Immunogen Full length human recombinant protein of human CLDN3 (NP_001297) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 1365
Target Species Human, Mouse, Rat, Zebrafish
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.