Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Claudin-4 Antibody (6E10), Novus Biologicals™
SDP

Catalog No. NB297669 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297669 100 μL
NB297668 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB297669 Supplier Novus Biologicals Supplier No. NBP326654100UL
Add to Cart
Add to Cart

Human Monoclonal Antibody

Claudin-4 Monoclonal antibody specifically detects Claudin-4 in Human samples. It is validated for ELISA

Specifications

Antigen Claudin-4
Applications ELISA
Classification Monoclonal
Clone 6E10
Conjugate Unconjugated
Dilution ELISA
Formulation 50% Glycerol, 0.01M PBS, PH 7.4
Gene Alias claudin 4, Clostridium perfringens enterotoxin receptor, Clostridium perfringens enterotoxin receptor 1, CPE-RCPETR, CPE-receptor, hCPE-R, WBSCR8CPERCPETR1claudin-4, Williams-Beuren syndrome chromosomal region 8 protein
Host Species Human
Immunogen Recombinant Human Claudin-4 protein [UniProt O14493]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 1364
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.