Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Claudin-5 Antibody (OTI1G4), PE/Atto594, Novus Biologicals™
SDP

Catalog No. N271329P594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N271329P594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N271329P594 Supplier Novus Biologicals Supplier No. NBP271329PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Claudin-5 Monoclonal specifically detects Claudin-5 in Human, Mouse, Rat, Porcine samples. It is validated for Flow Cytometry.

Specifications

Antigen Claudin-5
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1G4
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias AWALTransmembrane protein deleted in VCFS, BEC1, claudin 5, claudin-5, CPETRL1, TMVCFTMDVCF, transmembrane protein deleted in velocardiofacial syndrome
Gene Symbols CLDN5
Host Species Mouse
Immunogen Full length recombinant protein of human Claudin-5 (NP_003268) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 7122.0
Target Species Human, Mouse, Rat, Pig
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.