Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Claudin 5 Mouse anti-Human, Unconjugated, Clone: 2B12, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABT1528100UL
Description
Claudin 5 Monoclonal antibody specifically detects Claudin 5 in Human samples. It is validated for ELISA, Flow Cytometry, Inhibits Activity/Function, Western Blotting.
Specifications
Claudin 5 | |
Monoclonal | |
Unconjugated | |
Transmembrane protein deleted in VCFS;TMDVCF | |
Mouse | |
23.15 kDa calculated. | |
100 μL | |
Primary | |
Human | |
Purified |
ELISA, Flow Cytometry, Western Blot | |
2B12 | |
NP_003268 | |
CLDN5, AWAL, TMVCF | |
An artificial membrane protein consisting of the extracellular region of human Claudin 5. | |
Protein G Purified | |
Cell Structure | |
Clone 2B12 is a mouse monoclonal antibody that detects human Claudin 5. It targets an epitope within nine amino acids in the second extracellular domain. | |
Stable for 1 year at -20°C from date of receipt. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction