Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Claudin-6 Antibody (342927), HRP, Novus Biologicals™
SDP

Catalog No. NB348228 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB348228 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB348228 Supplier Novus Biologicals Supplier No. FAB3656H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Claudin-6 Monoclonal specifically detects Claudin-6 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry.

Specifications

Antigen Claudin-6
Applications CyTOF, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 342927
Conjugate HRP
Dilution Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation PBS
Gene Alias claudin 6, claudin-6, skullin
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Claudin-6, Met1-Val220, Accession # P56747
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9074
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.