Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Claudin-6 Antibody (9H8), Novus Biologicals™
SDP

Catalog No. NB297675 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297675 100 μL
NB297674 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB297675 Supplier Novus Biologicals Supplier No. NBP326651100UL
Add to Cart
Add to Cart

Human Monoclonal Antibody

Claudin-6 Monoclonal antibody specifically detects Claudin-6 in Human samples. It is validated for Flow Cytometry, ELISA

Specifications

Antigen Claudin-6
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 9H8
Conjugate Unconjugated
Dilution Flow Cytometry, ELISA
Formulation 50% Glycerol, 0.01M PBS, PH 7.4
Gene Alias claudin 6, claudin-6, skullin
Host Species Human
Immunogen Recombinant Human Claudin-6 protein [UniProt P56747]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9074
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.