Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Claudin-6 Antibody (9H8), Novus Biologicals™

Human Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Claudin-6 |
---|---|
Clone | 9H8 |
Dilution | Flow Cytometry, ELISA |
Applications | Flow Cytometry, ELISA |
Classification | Monoclonal |
Description
Claudin-6 Monoclonal antibody specifically detects Claudin-6 in Human samples. It is validated for Flow Cytometry, ELISASpecifications
Claudin-6 | |
Flow Cytometry, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
claudin 6, claudin-6, skullin | |
Recombinant Human Claudin-6 protein [UniProt P56747] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
9H8 | |
Flow Cytometry, ELISA | |
Unconjugated | |
Human | |
Stem Cell Markers | |
50% Glycerol, 0.01M PBS, PH 7.4 | |
9074 | |
IgG1 | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title