Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Claudin-8 Antibody (474210), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB368208 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB368208 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB368208 Supplier Novus Biologicals Supplier No. FAB5275L
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Claudin-8 Monoclonal antibody specifically detects Claudin-8 in Human samples. It is validated for Flow Cytometry, Flow Cytometry (Intracellular Staining), CyTOF

Specifications

Antigen Claudin-8
Applications Flow Cytometry, Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 474210
Conjugate DyLight 550
Dilution Flow Cytometry, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias claudin 8, human CLDN8 gene for claudin-810claudin-8
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human Claudin-8, Met1-Val225 (Ser151Pro), Accession # P56748
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9073
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.