Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLCA1 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001791 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001791 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001791 Supplier Novus Biologicals Supplier No. NB1001791
Add to Cart
Add to Cart

Goat Polyclonal Antibody

CLCA1 Polyclonal specifically detects CLCA1 in Human, Mouse samples. It is validated for Peptide ELISA, Western Blot.

Specifications

Antigen CLCA1
Applications Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:16000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_001276.1
Gene Alias CaCC, Calcium-activated chloride channel family member 1, Calcium-activated chloride channel protein 1, chloride channel accessory 1, chloride channel regulator, chloride channel regulator 1, chloride channel, calcium activated, family member 1, CLCRG1, FLJ95147, GOB5, hCaCC-1
Gene Symbols CLCA1
Host Species Goat
Immunogen Peptide with sequence C-TVTSKTNKDTSK corresponding to internal region according to NP_001276.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1179
Test Specificity No cross-reactivity expected with CLCA2 and 3.
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.