Learn More
Invitrogen™ CLCA1 Monoclonal Antibody (OTI8E7)

Description
CLCA1 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | CLCA1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI8E7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | CLCA1 |
| Gene Accession No. | A8K7I4 |
| Gene Alias | CACC; CACC1; CaCC-1; Calcium-activated chloride channel family member 1; calcium-activated chloride channel family member 3; Calcium-activated chloride channel protein 1; calcium-activated chloride channel regulator 1; calcium-dependent chloride channel-1; chloride channel accessory 1; chloride channel calcium activated 1; chloride channel calcium activated 2; chloride channel calcium activated 3; chloride channel regulator 1; chloride channel, calcium activated, family member 1; CLCA family member 1, chloride channel regulator; Clca1; Clca2; Clca3; CLCRG1; Gob5; gob-5; hCaCC-1; hCLCA1; mCLCA3; Protein gob-5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.