Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLCN3 Antibody (S258-5), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259379
Description
CLCN3 Monoclonal specifically detects CLCN3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
CLCN3 | |
Monoclonal | |
Unconjugated | |
PBS pH7.4, 50% glycerol with 0.09% Sodium Azide | |
CLCN3 | |
Synthetic peptide amino acids 98-115 (cytoplasmic N-terminus) of rat Clcn3 | |
100 μg | |
Primary | |
Detects 90kDa. Does not cross-react with Clcn4 or Clcn5 (based on KO validation results). | |
Store at -20C. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
S258-5 | |
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
chloride channel 3, Chloride channel protein 3, Chloride transporter ClC-3, ClC-3, CLC3, H(+)/Cl(-) exchange transporter 3 | |
Mouse | |
Protein G purified | |
RUO | |
1182 | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction