Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLCN3 Antibody (S258-5), Novus Biologicals™

Mouse Monoclonal Antibody
$560.50
Specifications
Antigen | CLCN3 |
---|---|
Clone | S258-5 |
Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
CLCN3 Monoclonal specifically detects CLCN3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
CLCN3 | |
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
1182 | |
Synthetic peptide amino acids 98-115 (cytoplasmic N-terminus) of rat Clcn3 | |
Primary | |
Detects 90kDa. Does not cross-react with Clcn4 or Clcn5 (based on KO validation results). |
S258-5 | |
Monoclonal | |
Purified | |
RUO | |
chloride channel 3, Chloride channel protein 3, Chloride transporter ClC-3, ClC-3, CLC3, H(+)/Cl(-) exchange transporter 3 | |
CLCN3 | |
IgG1 | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title