Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLCN6 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB096111 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB096111 100 μL
NB096112 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB096111 Supplier Novus Biologicals Supplier No. NBP304792100UL

Rabbit Polyclonal Antibody

CLCN6 Polyclonal specifically detects CLCN6 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CLCN6
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias chloride channel 6, Chloride channel protein 6, chloride transport protein 6, ClC-6, KIAA0046CLC-6
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-80 of human CLCN6 (NP_001277.1).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1185
Target Species Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.