Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLCN6 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00
Specifications
Antigen | CLCN6 |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
CLCN6 Polyclonal specifically detects CLCN6 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CLCN6 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
1185 | |
IgG | |
Affinity purified |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Mouse | |
chloride channel 6, Chloride channel protein 6, chloride transport protein 6, ClC-6, KIAA0046CLC-6 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-80 of human CLCN6 (NP_001277.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title