Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLE7 homolog Antibody (OTI2A4), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP271940UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271940UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271940UV Supplier Novus Biologicals Supplier No. NBP271940UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CLE7 homolog Monoclonal specifically detects CLE7 homolog in Human samples. It is validated for Western Blot.

Specifications

Antigen CLE7 homolog
Applications Western Blot
Classification Monoclonal
Clone OTI2A4
Conjugate DyLight 350
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CGI99, CGI-99, CLE, LCRP369, RLLM1
Gene Symbols RTRAF
Host Species Mouse
Immunogen Full length recombinant protein of human CLE7 homolog (NP_057123) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 51637.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.