Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLE7 homolog Antibody (OTI2A4), PE, Novus Biologicals™
SDP

Catalog No. NB288549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288549 Supplier Novus Biologicals Supplier No. NBP271940PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CLE7 homolog Monoclonal antibody specifically detects CLE7 homolog in Human samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen CLE7 homolog
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2A4
Conjugate PE
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias CGI99, CGI-99, CLE, LCRP369, RLLM1
Host Species Mouse
Immunogen Full length human recombinant protein of human C14orf166 (NP_057123) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 51637
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.