Learn More
Micronova™ Cleanroom NovaSqueegee™
SDP
A stainless steel squeegee tool with a replaceable rubber blade, designed to clean windows, screens, floors and walls in the cleanroom. Optional use with the SSU series universal handles allows extended reach.
$132.20 - $347.00
Specifications
For Use With (Equipment) NovaSqueegee
View More Specs
Products 9
Catalog Number Mfr. No. Product Type Material Length (English) Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Material Length (English) Price Quantity & Availability  
19-041-814A
SDP
View Documents
Micronova™
QDNS12
Squeegee Electro-polished Stainless Steel (304) 12 in.
Each for $305.00
Only null left
Add to Cart
19-041-814B
SDP
View Documents
Micronova™
QDNS18
Squeegee Electro-polished Stainless Steel (304) 18 in.
Each for $323.00
Only null left
Add to Cart
19-041-814C
SDP
View Documents
Micronova™
QDNS22
Squeegee Electro-polished Stainless Steel (304) 22 in.
Each for $326.00
Only null left
Add to Cart
19-041-803A
SDP
View Documents
Micronova™
RS12
Squeegee Blade Rubber 12 in.
Case of 12 for $132.20
Only null left
Add to Cart
19-041-803B
SDP
View Documents
Micronova™
RS18
Squeegee Blade Rubber 18 in.
Case of 12 for $347.00
Only null left
Add to Cart
19-041-803C
SDP
View Documents
Micronova™
RS22
Squeegee Blade Rubber 22 in.
Case of 12 for $182.10
Only null left
Add to Cart
19-041-802A
SDP
View Documents
Micronova™
NS12
Squeegee Blade Polyvinyl Acetate 12 in. Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
19-041-802B
SDP
View Documents
Micronova™
NS18
Squeegee Blade Polyvinyl Acetate 18 in. Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
19-041-802C
SDP
View Documents
Micronova™
NS22
Squeegee Blade Polyvinyl Acetate 22 in. Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description
Description
The Micronova Cleanroom NovaSqueegee™ is a stainless steel tool that comes standard with a replaceable rubber blade and features a Quick Disconnect handle attachment for optional use with the SSU series handles.
  • Stainless steel squeegee tool with a rubber blade
  • Designed to clean windows, screens, floors and walls in the cleanroom
  • Made with 304 electropolished stainless steel
  • Rubber replacement blades available
  • Available in 12, 18 and 22-inch (30, 45, 55 cm) sizes
  • Quick disconnect handle attachment
  • Optional use with SSU series Universal Handles
  • Entire unit is autoclavable
Specifications
Specifications
NovaSqueegee
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.